Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell AlphaTec™ BSL4 Orange Protective Suit with Attached PVC Safety Boots
Click to view available options
Size:
2X-Large (Suit), 12 (Boots)
2X-Large (Suit), 13 (Boots)
2X-Large (Suit), 14 (Boots)
Large (Suit), 10 (Boots)
Large (Suit), 11 (Boots)
Large (Suit), 12 (Boots)
Large (Suit), 9 (Boots)
Medium (Suit), 10 (Boots)
Medium (Suit), 7 (Boots)
Medium (Suit), 8 (Boots)
Medium (Suit), 9 (Boots)
Small (Suit), 6 (Boots)
Small (Suit), 7 (Boots)
Small (Suit), 8 (Boots)
X-Large (Suit), 11 (Boots)
X-Large (Suit), 12 (Boots)
X-Large (Suit), 13 (Boots)
X-Large (Suit), 14 (Boots)
Description
- Suitable for Level 4 Biological Safety Laboratories
- Provides protection against chemicals in liquid, aerosol and solid form
- Protects against infectious agents, i.e. bacteria, virus and fungi, and against radioactive particles
- Designed for use with an external air source, which allows longer working hours and provides improved wearer comfort
- Made from a strong yet soft and durable material

Specifications
Specifications
Color | Orange |
For Use With (Application) | Biosafety Labs Levels 1-4, Centers for Disease Control and Prevention, Life Sciences, Chemical, Mining, Fire and Rescue, Oil and Gas |
Material | Polyester |
Size | Medium (Suit), 9 (Boots) |
Closure Type | Heavy-duty vapor-protective zipper with D-rings |
Disposable | Reusable |
Product Type | Chemical Protective Suits |
Seam Type | Stitched and welded |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction