Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 87-089 Yellow Lightweight Latex Gloves

Catalog No. 19010224 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5
7.5
8.5
9.5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-224 9.5
19-010-221 6.5
19-010-222 7.5
19-010-223 8.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-010-224 Supplier Ansell Supplier No. 6615
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Natural rubber latex gloves are flock lined and lightweight with tear resistant beaded cuffs.

  • Natural rubber latex
  • Abrasion/tear resistant
  • 100% cotton flock lining
  • Beaded cuff for easy donning
  • Ideal for agriculture, food processing or chemical applications that are low hazard risk
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE0493, EN420:2003 + A1:2009
Chlorinated Yes
AQL 0.65
Color Yellow
Latex Free No
Material Latex
Size 9.5
Thickness (English) 17 mil
Thickness (Metric) 0.43 mm
EN388 Standard EN 388:2010, EN 388:2010X
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Beaded
Grip Design Reversed Lozenge
Liner Material Cotton Flock
Lining Lined
Model G12Y 9.5 113520
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.