Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 2000 Comfort Bound Coveralls with Hood, Model 129

Catalog No. 17999890
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls protective barrier shields hood, arms, legs, and front torso against particulates, liquids, and light chemical splash. Three-piece hood ensures adequate coverage and finger loops improve comfort and usability. Previously known as MICROCHEM™ by AlphaTec 2000 Comfort.

  • Arms, legs, and front torso in 2000 fabric
  • Protects against liquid, light chemical resistance,and particulate biological agents
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free
  • Anti-static, tested according to EN1149-5 and AATCC76
  • Elastic ankles and wrists
  • 3-piece hood, 2-way front zipper with storm flap
  • Bound seams, finger loops
  • Ideal for pharmaceutical industries, cleanrooms, paint spraying, veterinary services, pest control
  • Available in Small through 5X-Large
  • Certifications EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Laminate, SMS
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Chemicals
Back Style Breathable SMS Back Panel
Certifications/Compliance AATTC 76 Compliant
Certifications/Compliance (Europe) EN 1149-5, CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
Latex Free Yes
Model WH20-B-92-129
For Use With (Application) Automotive, Life Sciences, Metal Fabrication, Veterinary, Maintenance, Cleaning, Production, Food Processing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.