Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALPI Mouse anti-Bovine, Clone: P4A10E4, Abnova™

Catalog No. 89035386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-386 Supplier Abnova Corporation Supplier No. MAB13979

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native bovine ALPI.

  • High specificity
  • High reactivity

Specifications

Antigen ALPI
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone P4A10E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native bovine ALPI.
Dilution ELISA Immunohistochemistry Immunoprecipitation Radioimmunoassay The optimal working dilution should be determined by the end user.
Formulation In PBS (1 mg/mL BSA, 0.05% sodium azide).
Gene Alias IAP
Gene Symbols ALPI
Host Species Mouse
Immunogen Native purified ALPI from bovine calf intestine.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280993
Target Species Bovine
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.