Learn More
ALPI Mouse anti-Human, Clone: OTI3C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809178

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.Specifications
ALPI | |
Monoclonal | |
Unconjugated | |
ALPI | |
IAP | |
Mouse | |
Affinity Chromatography | |
RUO | |
248 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09923 | |
ALPI | |
Full length human recombit protein of human ALPI produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.