Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101243 Supplier Invitrogen™ Supplier No. PA5101243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ALPK1.

Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALPK1
Gene Accession No. Q96QP1, Q9CXB8
Gene Alias 8430410J10Rik; alpha kinase 1; alpha-kinase 1; alpha-protein kinase 1; Alpk1; Chromosome 4 kinase; KIAA1527; LAK; lymphocyte alpha-kinase; lymphocyte alpha-protein kinase; RGD1304613
Gene Symbols ALPK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALPK1(Accession Q96QP1), corresponding to amino acid residues G894-G944.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71481, 80216
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.