Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120710 Supplier Invitrogen™ Supplier No. PA5120710
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, SW620. Immunogen sequence: RDDEPVTGKQ ELHSFVKAAF GLTTVHRRLH GETGTVHAAS QLCKEAMGKL YNFSTSSRSQ DREALSQEVM SVIAQVKEHL QVQSFSNVDD RSYVPESFEC RLDKLILHGQ GDFQKILDTY SQHHTSVCEV FESDCGNNKN EQKDAKTGVC ITALKTEIKN I.

Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ALPK1
Gene Accession No. Q96QP1
Gene Alias 8430410J10Rik; alpha kinase 1; alpha-kinase 1; alpha-protein kinase 1; Alpk1; Chromosome 4 kinase; KIAA1527; LAK; lymphocyte alpha-kinase; lymphocyte alpha-protein kinase; RGD1304613
Gene Symbols ALPK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-500 of human ALPK1 (NP_001095876.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80216
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.