Learn More
Invitrogen™ ALPK2 Monoclonal Antibody (4E5)

Description
Peptide Sequence: DGTSSVTEQG RYKLPTAPEA AENDYPGIQG ETRDSHQARE EFASDNLLNM DESVRETEMK LLSGESENSG MSQCWETAAD KRVGGKDLWS KRGSRKSARV RQPGMKGNP.
Specifications
Specifications
| Antigen | ALPK2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4E5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ALPK2 |
| Gene Accession No. | Q86TB3 |
| Gene Alias | alpha kinase 2; alpha-kinase 2; alpha-protein kinase 2; Alpk2; Gm549; HAK; heart alpha-kinase; heart alpha-protein kinase; RGD1564431; RNF18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.