Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556263 Supplier Invitrogen™ Supplier No. PA556263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEAPFTGTTT ISFSNLGGVH KENASLAQHS EVKPCTCGPQ HEEKQDRDGN IPDNFREDLK YEQSISEAND ETMSPGVFSR HLPKDARAD Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 35%.

Protein kinase that recognizes phosphorylation sites in which the surrounding peptides have an alpha-helical conformation (PubMed:10021370). Regulates cardiac development and cardiomyocyte differentiation by negatively regulating Wnt/beta-catenin signaling (PubMed:29888752). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPK2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALPK2
Gene Accession No. Q86TB3
Gene Alias alpha kinase 2; alpha-kinase 2; alpha-protein kinase 2; Alpk2; Gm549; HAK; heart alpha-kinase; heart alpha-protein kinase; RGD1564431; RNF18
Gene Symbols ALPK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALPK2. Recombinant protein control fragment (Product #RP-95080).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.