Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555344 Supplier Invitrogen™ Supplier No. PA555344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISSLSQAPEC GAQSLGKAPP QASVQVPTPP ARRRHGTRDS TLQGQAGHRT PGEVLECQTT TAPTMSASSS SDVASIGVST SGSQGIIEPM DMETQE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 72%.

Predicted to enable ATP binding activity; protein serine kinase activity; and protein serine/threonine kinase activity. Predicted to be involved in cardiac muscle cell development. Predicted to be active in nucleus. Implicated in hypertrophic cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPK3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALPK3
Gene Accession No. Q96L96
Gene Alias alpha kinase 3; alpha-kinase 3; alpha-protein kinase 3; ALPK3; AW319487; D330016D04; KIAA1330; MAK; Midori; mKIAA1330; muscle alpha-kinase; muscle alpha-protein kinase; myocyte induction differentiation originator; myocytic induction/differentiation originator
Gene Symbols ALPK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAK. Recombinant protein control fragment (Product #RP-95065).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.