Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALPK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103350
Description
Antibody detects endogenous levels of total ALPK3.
Predicted to enable ATP binding activity; protein serine kinase activity; and protein serine/threonine kinase activity. Predicted to be involved in cardiac muscle cell development. Predicted to be active in nucleus. Implicated in hypertrophic cardiomyopathy.
Specifications
ALPK3 | |
Polyclonal | |
Unconjugated | |
ALPK3 | |
alpha kinase 3; alpha-kinase 3; alpha-protein kinase 3; ALPK3; AW319487; D330016D04; KIAA1330; MAK; Midori; mKIAA1330; muscle alpha-kinase; muscle alpha-protein kinase; myocyte induction differentiation originator; myocytic induction/differentiation originator | |
Rabbit | |
Affinity chromatography | |
RUO | |
116904, 57538 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q924C5, Q96L96 | |
ALPK3 | |
A synthesized peptide derived from human ALPK3(Accession Q96L96), corresponding to amino acid residues L1824-P1874. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction