Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578763
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578763 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578763 Supplier Invitrogen™ Supplier No. PA578763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell. IHC: human parotid acinar cell carcinoma tissue.

Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism's environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ALPL
Gene Accession No. P05186
Gene Alias Akp2; Akp-2; Alkaline phosphatase 2; alkaline phosphatase 2, liver; alkaline phosphatase liver/bone/kidney isozyme; alkaline phosphatase precursor (EC 3.1.3.2); alkaline phosphatase tissue non-specific isoform; alkaline phosphatase tissue non-specific isoform/TNS-AP; alkaline phosphatase, biomineralization associated; alkaline phosphatase, intestinal; alkaline phosphatase, liver/bone/kidney; alkaline phosphatase, tissue-nonspecific isozyme; alkaline phosphomonoesterase; ALPI; Alpl; APTNAP; AP-TNAP; FLJ40094; FLJ93059; glycerophosphatase; HOPS; IAP; intestinal alkaline phosphatase; intestinal-type alkaline phosphatase; Kasahara isozyme; liver/bone/kidney isozyme; liver/bone/kidney-type alkaline phosphatase; MGC161443; MGC167935; PHOA; tissue-nonspecific alkaline phosphatase; tissue-nonspecific alkaline phosphatase precursor; tissue-nonspecific ALP; Tissue-nonspecific ALP alkaline phosphatase; TNAP; TNSALP; TNS-AP
Gene Symbols ALPL
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human Alkaline Phosphatase (21-35aa,EKEKDPKYWRDQAQE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 249
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.