Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALPP Rabbit anti-Human, Clone:RM317, Abnova™

Catalog No. 89333298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-298 Supplier Abnova Corporation Supplier No. MAB16942
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against huamn ALPP.

There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The first three are located together on chromosome 2 while the tissue non-specific form is located on chromosome 1. The product of this gene is a membrane bound glycosylated enzyme, also referred to as the heat stable form, that is expressed primarily in the placenta although it is closely related to the intestinal form of the enzyme as well as to the placental-like form. The coding sequence for this form of alkaline phosphatase is unique in that the 3′ untranslated region contains multiple copies of an Alu family repeat. In addition, this gene is polymorphic and three common alleles (type 1, type 2 and type 3) for this form of alkaline phosphatase have been well characterized. [provided by RefSeq]

Specifications

Antigen ALPP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM317
Conjugate Unconjugated
Description alkaline phosphatase, placental (Regan isozyme)
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1000)
Western Blot (1:1000-2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene ALPP
Gene Alias ALP, FLJ61142, PALP, PLAP
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human ALPP.
Quantity 100 μL
Gene ID (Entrez) 250
Test Specificity This antibody reacts to human PLAP (Placental alkaline phosphatase).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.