Learn More
Invitrogen™ ALPPL2 Polyclonal Antibody

Description
Recommended positive controls: A549. Predicted reactivity: Human (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ALPPL2 |
| Applications | Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M glycine, 0.1M tris with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ALPG |
| Gene Accession No. | P10696 |
| Gene Alias | Akp5; alkaline phosphatase 5; Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; alkaline phosphatase, placental like 2; alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; Alpg; ALPPL; ALPPL2; C77216; D1Ertd816e; EAP; Embryonic alkaline phosphatase; embryonic-type alkaline phosphatase; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; RGD1565100; testicular and thymus alkaline phosphatase |
| Gene Symbols | ALPG, ALPPL2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.