Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559367 Supplier Invitrogen™ Supplier No. PA559367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNLTRLPDGV PGGTQALWLD GNNLSSVPPA AFQNLSSLGF LNLQGGQLGS LEPQALLGLE NLCHLHLERN QLRSLALGTF AHTPALASLG LSNNRLSR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 74%.

The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGFALS
Gene Accession No. P35858
Gene Alias ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111
Gene Symbols IGFALS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALS. Recombinant protein control fragment (Product #RP-98358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.