Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587723 Supplier Invitrogen™ Supplier No. PA587723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCLGRIRPHT FTGLSGLRRL FLKDNGLVGI EEQSLWGLAE LLELDLTSNQ LTHLPHRLFQ GLGKLEYLLL SRNRLAELPA DALGPLQRAF WLDVSHNRLE ALPNSLLAPL GRLRYLSLRN NSLRTFTPQP PGLERLWLEG NPWDCGCPLK ALRDFALQNP SAVPRFVQAI CEGDDCQPPA YTYNNITCAS PPEVVGLDLR DLSEAHFAPC; Positive Samples: Mouse lung, Rat liver; Cellular Location: extracellular space.

The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IGFALS
Gene Accession No. P35859, P70389
Gene Alias ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111
Gene Symbols IGFALS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 396-605 of human IGFALS (NP_0049611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16005, 79438
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.