Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Als2 Antibody, Novus Biologicals™
SDP

Catalog No. NB100885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100885 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100885 Supplier Novus Biologicals Supplier No. NB100885
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Als2 Polyclonal specifically detects Als2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Als2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q96Q42
Gene Alias ALS2CR6, alsin, ALSJ, amyotrophic lateral sclerosis 2 (juvenile), amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 6, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein, Amyotrophic lateral sclerosis 2 protein, FLJ31851, IAHSP, KIAA1563, MGC87187, PLSJ
Gene Symbols ALS2
Host Species Goat
Immunogen Peptide with sequence LKACYYQIQREKLN corresponding to C-Terminus according to NP_065970.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57679
Test Specificity Expected from sequence similarity: Rat, Dog, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.