Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2 Mouse anti-Mouse, Human, Bovine, Rat, Clone: N-alsin-24, MilliporeSigma™

Catalog No. MABN262825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN262825 25 μg
MABN2628100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN262825 Supplier MilliporeSigma Supplier No. MABN262825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALS2 Monoclonal antibody specifically detects ALS2 in Mouse, Human, Bovine, Rat samples. It is validated for ELISA, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone N-alsin-24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to the first 364 amino acids from the N-terminal region of murine Alsin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Human, Bovine, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.