Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561704 Supplier Invitrogen™ Supplier No. PA561704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDPREKLEVL ERTYGEIEGT VSRVLGREYK LPMDDLLPLL IYVVSRARIQ HLGAEIHLIR DMMDPNHTGG LYDFLLTALE S Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 95%.

Acts as a guanine nucleotide exchange factor (GEF) for Rab5 GTPase. Regulates the ALS2-mediated endosome dynamics. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALS2CL
Gene Accession No. Q60I27
Gene Alias ALS2 C-terminal like; ALS2 C-terminal-like protein; ALS2CL; RN49018
Gene Symbols ALS2CL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALS2CL. Recombinant protein control fragment (Product #RP-99934).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.