Learn More
Invitrogen™ ALS2CR1 Monoclonal Antibody (OTI1F6)

Description
ALS2CR1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | ALS2CR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1F6 |
| Concentration | 0.85 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NIF3L1 |
| Gene Accession No. | Q9GZT8 |
| Gene Alias | 1110030G24Rik; ALS2CR1; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 1; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein; CALS-7; GTP cyclohydrolase I; MDS015; My018; Ngg1 interacting factor 3 homolog-like 1; NGG1 interacting factor 3 like 1; Ngg1 interacting factor 3-like 1; Ngg1 interacting factor 3-like 1 (S. pombe); NIF3 (Ngg1 interacting factor 3, S.pombe homolog)-like 1; NIF3 NGG1 interacting factor 3-like 1; NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae); Nif3l1; NIF3-like protein 1; OTTHUMP00000206190; OTTHUMP00000206191; OTTHUMP00000206192; putative GTP cyclohydrolase 1 type 2 Nif3l1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.