Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111073 Supplier Invitrogen™ Supplier No. PA5111073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLSSLNDFAS LSFAESWDNV GLLVEPSPPH TVNTLFLTND LTEEVMEEVL QKKADLILSY HPPIFRPMKR ITWNTWKERL VIRALE Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 93%.

ALS2CR1 may function as a transcriptional corepressor through its interaction with COPS2, negatively regulating the expression of genes involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIF3L1
Gene Accession No. Q9GZT8
Gene Alias 1110030G24Rik; ALS2CR1; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 1; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein; CALS-7; GTP cyclohydrolase I; MDS015; My018; Ngg1 interacting factor 3 homolog-like 1; NGG1 interacting factor 3 like 1; Ngg1 interacting factor 3-like 1; Ngg1 interacting factor 3-like 1 (S. pombe); NIF3 (Ngg1 interacting factor 3, S.pombe homolog)-like 1; NIF3 NGG1 interacting factor 3-like 1; NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae); Nif3l1; NIF3-like protein 1; OTTHUMP00000206190; OTTHUMP00000206191; OTTHUMP00000206192; putative GTP cyclohydrolase 1 type 2 Nif3l1
Gene Symbols NIF3L1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ALS2CR1. Recombinant protein control fragment (Product #RP-96226).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60491
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.