Learn More
ALS2CR11 Mouse anti-Human, Clone: OTI7A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811585

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ALS2CR11 has 2 isoforms of the human protein that are produced by alternative splicing. An autosomal recessive form of juvenile amyotrophic lateral sclerosis was originally mapped to a region of chromosome 2 that includes this gene. The encoded protein contains a calcium-dependent membrane targeting C2 domain. This domain is often found in proteins that are involved in membrane trafficking and signal transduction.Specifications
ALS2CR11 | |
Monoclonal | |
Unconjugated | |
C2CD6 | |
ALS2CR11 | |
Mouse | |
Affinity Chromatography | |
RUO | |
151254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53TS8 | |
C2CD6 | |
Human recombit protein fragment corresponding to amino acids 307-623 of human ALS2CR11 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.