Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CR11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549977 Supplier Invitrogen™ Supplier No. PA549977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ALS2CR11 protein.

ALS2CR11 has 2 isoforms of the human protein that are produced by alternative splicing. An autosomal recessive form of juvenile amyotrophic lateral sclerosis was originally mapped to a region of chromosome 2 that includes this gene. The encoded protein contains a calcium-dependent membrane targeting C2 domain. This domain is often found in proteins that are involved in membrane trafficking and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C2CD6
Gene Accession No. Q53TS8
Gene Alias 1700052H20Rik; 4930408G06Rik; Als2cr11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human); amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 11; C2 calcium-dependent domain-containing protein 6; C2CD6; testicular tissue protein Li 16
Gene Symbols Als2cr11, C2CD6
Host Species Rabbit
Immunogen Synthesized peptide derived from C-terminal of human ALS2CR11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151254, 73463
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.