Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALS2CR12 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305958MFV610
Description
ALS2CR12 Polyclonal antibody specifically detects ALS2CR12 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
ALS2CR12 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12 | |
Produced in rabbits immunized with E. coli-derived Human ALS2CR12 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
130540 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction