Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557399 Supplier Invitrogen™ Supplier No. PA557399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSARQEETNK SFYEVINVSP GYQLVRNREQ ISVTLGDEMF DRKKRWESEI PDKGRFSRTN IISDLEEQIS ELTAIIEQMN R Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 73%.

ALS2CR12 is a protein coding gene and gene ontology (GO) annotation includes cytoplasm; outer dense fiber; sperm fibrous sheath; sperm flagellum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR12
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALS2CR12
Gene Accession No. Q96Q35
Gene Alias ALS2CR12; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 12; amyotrophic lateral sclerosis 2 chromosome region, candidate 12; FLACC1; Flagellum-associated coiled-coil domain-containing protein 1
Gene Symbols ALS2CR12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALS2CR12. Recombinant protein control fragment (Product #RP-96156).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.