Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598776
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598776 Supplier Thermo Scientific Supplier No. PA598776
Add to Cart
Add to Cart

ALS2CR12 Polyclonal Antibody

ALS2CR12 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR12
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ALS2CR12
Gene Accession No. Q96Q35
Gene Alias ALS2CR12; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 12; amyotrophic lateral sclerosis 2 chromosome region, candidate 12; FLACC1; Flagellum-associated coiled-coil domain-containing protein 1
Gene Symbols ALS2CR12
Host Species Rabbit
Immunogen Recombinant Human Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein (26-76AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130540
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.