Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531636 Supplier Invitrogen™ Supplier No. PA531636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ALS2CR12 is a protein coding gene and gene ontology (GO) annotation includes cytoplasm; outer dense fiber; sperm fibrous sheath; sperm flagellum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ALS2CR12
Gene Accession No. Q96Q35
Gene Alias ALS2CR12; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 12; amyotrophic lateral sclerosis 2 chromosome region, candidate 12; FLACC1; Flagellum-associated coiled-coil domain-containing protein 1
Gene Symbols ALS2CR12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALS2CR12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.