Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALS2CR2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100442APC
Description
ALS2CR2 Polyclonal antibody specifically detects ALS2CR2 in Mouse samples. It is validated for Western BlotSpecifications
| ALS2CR2 | |
| Polyclonal | |
| Western Blot | |
| ALS2CR2candidate 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein, CALS-21MGC102916, ILP-interacting protein, ILPIPSTRAD beta, Pseudokinase ALS2CR2, STE20-related kinase adaptor beta | |
| Residues 399-418 (SPWSELEFQFPDDKDPVWEF) of the mouse PAPK protein | |
| 0.1 mL | |
| Apoptosis, Protein Kinase, Signal Transduction | |
| 55437 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction