Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALS2CR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14314
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14314 Supplier Invitrogen™ Supplier No. PA14314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4314 detects the PAPK protein in mouse samples. PA1-4314 has been successfully used in Western blot procedures. The PA1-4314 immunizing peptide corresponds to amino acid residues 399-418 of the mouse PAPK protein.

This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene STRADB
Gene Accession No. Q8K4T3
Gene Alias AA792893; Als2cr2; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 2; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein homolog; B830008M19; CALS-21; D1Ucla2; ILP-interacting protein; ILP-interacting protein homolog; ILP-interacting protein ILPIPA; ILPIP; ILPIPA; MGC102916; Papk; polyploidy associated protein kinase; polyploidy associated protein kinase PAPK-A; polyploidy-associated protein kinase; PRO1038; pseudokinase ALS2CR2; RGD1559449; STE20 related adaptor beta; STE20-related kinase adapter protein beta; STE20-related kinase adaptor beta; STRAD beta; Stradb; Syradb
Gene Symbols STRADB
Host Species Rabbit
Immunogen Synthetic peptide: S(399) P W S E L E F Q F P D D K D P V W E F(418).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227154
Target Species Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.