Learn More
Invitrogen™ ALS2CR2 Polyclonal Antibody

Description
Immunogen sequence: FSPAFFSLVQ LCLQQDPEKR PSASSLLSHV FFKQMKEESQ DSILSLLPPA YNKPSISLPP VLPWTEPECD FPDE Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 88%.
Specifications
Specifications
| Antigen | ALS2CR2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.30 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | STRADB |
| Gene Accession No. | Q8K4T3, Q9C0K7 |
| Gene Alias | AA792893; Als2cr2; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 2; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein homolog; B830008M19; CALS-21; D1Ucla2; ILP-interacting protein; ILP-interacting protein homolog; ILP-interacting protein ILPIPA; ILPIP; ILPIPA; MGC102916; Papk; polyploidy associated protein kinase; polyploidy associated protein kinase PAPK-A; polyploidy-associated protein kinase; PRO1038; pseudokinase ALS2CR2; RGD1559449; STE20 related adaptor beta; STE20-related kinase adapter protein beta; STE20-related kinase adaptor beta; STRAD beta; Stradb; Syradb |
| Gene Symbols | STRADB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.