Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALS2CR3 Antibody (S390-43), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | ALS2CR3 |
---|---|
Clone | S390-43 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ALS2CR3 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Human, Rat | |
66008 | |
Fusion protein amino acids 757-914 (C-terminus) of human TRAK2 | |
Primary | |
Detects 100kDa. Does not cross-react with TRAK1. |
S390-43 | |
Monoclonal | |
Purified | |
RUO | |
ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2 | |
TRAK2 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title