Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7925 Supplier Bioss Supplier No. BS7684R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARF (calcium-response factor), also known as ALS2CR8 (amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein) or NYD-SP24, is a 725 amino acid nuclear protein. Expressed in a wide variety of tissues with highest expression in the hippocampus, CARF is thought to be a transcription factor that associates with the p53 tumor suppression pathway. CARF cooperates, co-localizes and is co-regulated with ARF, an ADP-ribosylation factor, and, through this interaction, helps to mediate ARF-p53-induced apoptotic signaling. This apoptotic pathway is implicated in cell cycle control, proper cellular development, response to DNA damage and the aging process, suggesting that CARF participates in various events throughout the cell.

Specifications

Antigen ALS2CR8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CARF
Gene Accession No. Q8N187
Gene Alias ALS2CR8; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein homolog; calcium response factor; calcium responsive transcription factor; Calcium-response factor; calcium-response factor CaRF; Calcium-responsive transcription factor; Carf; ECBRC-FC1; ECBRC-FC2; NYD-SP24; Testis development protein NYD-SP24
Gene Symbols CARF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ALS2CR8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79800
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.