Learn More
Invitrogen™ Alsin Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000. Preliminary analysis showed no signal in Western Blot.
Specifications
Specifications
| Antigen | Alsin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ALS2 |
| Gene Accession No. | Q96Q42 |
| Gene Alias | 3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ |
| Gene Symbols | ALS2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.