Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alsin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561254 Supplier Invitrogen™ Supplier No. PA561254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQPGLYYSGR QDPTEGDNLP ENHSGSKTPV LLSCSKLGYI SRVTAGKDSY LALVDKNIMG YIASLHELAT TERRFYSKLS D Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alsin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALS2
Gene Accession No. Q96Q42
Gene Alias 3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ
Gene Symbols ALS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Alsin. Recombinant protein control fragment (Product #RP-95946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57679
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.