Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Alsin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103595
Description
Antibody detects endogenous levels of total ALS2.
The function of this protein remains unknown.
Specifications
Alsin | |
Polyclonal | |
Unconjugated | |
ALS2 | |
3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ | |
Rabbit | |
Affinity chromatography | |
RUO | |
363235, 57679, 74018 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P0C5Y8, Q920R0, Q96Q42 | |
ALS2 | |
A synthesized peptide derived from human ALS2(Accession Q96Q42), corresponding to amino acid residues L204-S254. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction