Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers

Catalog No. p-4530856
Click to view available options
Type:
Standard Plug (Factory Installed)
Twist Lock (Factory Installed)
Twist Lock (Field Installed)
Plug Type:
US NEMA 6-15 P
US NEMA 6-20 P
US NEMA L5-20 P
US NEMA L6-15 P
Amperage:
15 A
20 A
Voltage:
120 V
230 V

Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers.

Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers. Options include an array of standard and special plugs for North America and international markets.

TRUSTED_SUSTAINABILITY

Specifications

Type Twist Lock (Factory Installed)
For Use With (Equipment) Thermo Scientific™ TDE Series, HERAfreeze HFU B, Revco™ ExF/DxF, Thermo Scientific™ TSE Series
Amperage 20 A
Plug Type US NEMA L5-20 P
Voltage 120 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.