Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alumina, 100-200 Mesh, Reagents

Catalog No. 01337553
Click to view available options
Quantity:
2000 g
5 lb.

Alumina, 100 to 200 Mesh, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym Aluminum oxide
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2977°C
Color White to Off-White
Physical Form Solid
Quantity 2000 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.8
Chemical Name or Material Alumina, 100-200 Mesh
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.