Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum, 99.5%, pieces (2-10 mm)

Catalog No. AC300210010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300210010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC300210010 Supplier Thermo Scientific Chemicals Supplier No. 300210010
Add to Cart

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Gray
Melting Point 660.0°C
Boiling Point 2327.0°C
Density 2.7000g/mL
Quantity 1 kg
Packaging Glass bottle
Form Shot
Assay Percent Range 99.5%
Fieser 02,19; 04,9; 12,486; 16,337
IUPAC Name aluminum
Formula Weight 26.97
Percent Purity 99.5%
Chemical Name or Material Aluminum, 99.5%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement AIR SENSITIVE
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.