Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum acetylacetonate, 97%

Catalog No. p-3756523
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle

Catalog No. AC196970050

Supplier: Thermo Scientific Chemicals 196970050

Only null left
Add to Cart

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym aluminum 2, 4-pentanedionate
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color White to Cream
Boiling Point 315.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Glass bottle
Linear Formula (CH3COCH2COCH2)3Al
Quantity 5 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: 4.26 g/100g alcohol (20°C), 36.45 g/100g benzene (20°C), insoluble in petroleum ether
Formula Weight 324.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Aluminum acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
EINECSNumber 237-741-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.