Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Lewis acid

Specifications
Specifications
Melting Point | 194.0°C |
Color | Gray or White to Yellow |
Physical Form | Powder |
Quantity | 2.5 kg |
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | AlCl3 |
Fieser | 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15 |
Merck Index | 15, 333 |
Solubility Information | Solubility in water: reacts. Other solubilities: freely miscible with benzene,nitrobenzene,,benzophenone,carbon tetrachloride and chloroform |
Formula Weight | 133.34 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Reacts violently with water. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately |
EINECSNumber | 231-208-1 |
RTECSNumber | BD0525000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction