Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum chloride hexahydrate, 99%, pure

Catalog No. p-3734618
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Quantity 500 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AlCl3·6H2O
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15
Merck Index 15,333
Solubility Information Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene
IUPAC Name aluminium(3+) hexahydrate trichloride
Formula Weight 241.43
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Specific Gravity 2.39
Density 2.3900g/mL
Chemical Name or Material Aluminum chloride hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medical advice/attention.
Wash face,hands and any ex

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.