Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis

Catalog No. p-2691013
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 7784-24-9 | AlKO8S2·12H2O | 474.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlKO8S2·12H2O
Molecular Weight (g/mol) 474.39
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92.0°C
Color White
pH 3 to 3.5 (10% soln. at 20°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99.5% min. (Complexometry)
Linear Formula AlK(SO4)2·12H2O
Merck Index 15, 354
Solubility Information Solubility in water: 140g/L (20°C). Other solubilities: freely soluble in glycerol, insoluble in alcohol
Formula Weight 474.39
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Aluminum potassium sulfate dodecahydrate, p.a.
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.