Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium sulfate octadecahydrate, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
7784-31-8
Al2H36O30S3
666.401
MFCD00149136
AMVQGJHFDJVOOB-UHFFFAOYSA-H
aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminium sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminium sulfate-water 1/18
22377415
CHEBI:74779
dialuminum;trisulfate;octadecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
7784-31-8
Al2H36O30S3
666.401
MFCD00149136
AMVQGJHFDJVOOB-UHFFFAOYSA-H
aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminium sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminium sulfate-water 1/18
22377415
CHEBI:74779
dialuminum;trisulfate;octadecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Linear Formula Al2(SO4)3·18H2O
Ammonium 30ppm max.
Arsenic (As) 5ppm max.
Formula Weight 666.4
Physical Form Liquid
Percent Purity 98.0 to 110.0% (Complexometry)
Grade Extra Pure
Chloride 50ppm max.
Heavy Metals (as Pb) 20ppm max.
Insoluble Matter 0.01% max.
Iron (Fe) 50ppm max.
Packaging Plastic drum
Solubility Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Substances not ppt by NH3 0.2% max. (as SO4)
Color Colorless to Yellow
Melting Point 86.0°C
pH 3 to 4 (2% soln. at 20°C)
Quantity 10kg
Chemical Name or Material Aluminium sulfate octadecahydrate
Show More Show Less

GHS P Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : WS5697000