Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium tri-sec-butoxide, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
2269-22-9
C12H27AlO3
246.33
MFCD00009327
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum sec-butoxide,aluminum tri-sec-butoxide,2-butanol, aluminum salt,aluminum sec-butylate,aluminum butan-2-olate,aluminium tri-s-butoxide,2-butanol, aluminum salt 3:1,aluminium tri-sec-butanolate,al o-sec-bu 3,aluminiumtri-sec-butoxide
50931103
aluminum;butan-2-ol
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
2269-22-9
C12H27AlO3
246.33
MFCD00009327
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum sec-butoxide,aluminum tri-sec-butoxide,2-butanol, aluminum salt,aluminum sec-butylate,aluminum butan-2-olate,aluminium tri-s-butoxide,2-butanol, aluminum salt 3:1,aluminium tri-sec-butanolate,al o-sec-bu 3,aluminiumtri-sec-butoxide
50931103
aluminum;butan-2-ol
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
Lewis acid
Specific Gravity 0.96
Viscosity 1800 mPa.s (20°C)
Refractive Index 1.4380 to 1.4400
Quantity 100g
Density 0.9600g/mL
Boiling Point 200.0°C to 206.0°C (30.0mmHg)
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 27°C
Assay Percent Range 96% min. (ex Al)
Linear Formula [C2H5CH(CH3)O]3Al
Beilstein 01,IV,1567
Solubility Information Solubility in water: decomposes
Formula Weight 246.33
Physical Form Oily Liquid
Percent Purity 97%
Chemical Name or Material Aluminium tri-sec-butoxide
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye

GHS Signal Word: Warning

EINECSNumber : 218-871-2