Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium trifluoromethanesulfonate, 99%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Plastic bottle
74974-61-1
C3AlF9O9S3
474.19
MFCD00143596
FKOASGGZYSYPBI-UHFFFAOYSA-K
aluminum trifluoromethanesulfonate,aluminum triflate,aluminium triflate,aluminum tris trifluoromethanesulfonate,aluminium trifluoromethanesulphonate,aluminium 3+ tritriflate,aluminium trifluoromethanesulfonate,aluminum iii triflate,aluminium iii triflate,aluminum tris fluoranyl methanesulfonate
2737634
aluminum;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3]
74974-61-1
C3AlF9O9S3
474.19
MFCD00143596
FKOASGGZYSYPBI-UHFFFAOYSA-K
aluminum trifluoromethanesulfonate,aluminum triflate,aluminium triflate,aluminum tris trifluoromethanesulfonate,aluminium trifluoromethanesulphonate,aluminium 3+ tritriflate,aluminium trifluoromethanesulfonate,aluminum iii triflate,aluminium iii triflate,aluminum tris fluoranyl methanesulfonate
2737634
aluminum;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3]
Assay Percent Range 98.5% min. (Complexometry)
Solubility Information Solubility in water: insoluble
Formula Weight 474.19
Physical Form Powder
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: insoluble.
Color White
Melting Point 300.0°C
Quantity 10g
Chemical Name or Material Aluminium trifluoromethanesulfonate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger