Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder, Approx. 30%, Spectrum™ Chemical
SDP

Catalog No. 18600461
Click to view available options
Quantity:
125 g
500 g
2.5 kg
150 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

AL223, C2H5O4Al·H3BO3

Spectrum™ Chemical Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7360-44-3, 10043-35-3

Specifications

CAS Max % 30%, 70%
Quantity 500 g
Percent Purity ∼30%
Grade Reagent
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.