Aluminum ammonium sulfate dodecahydrate, White crystalline, Alfa Aesar™

Manufacturer: Alfa Aesar 13802A3

Quantity 2kg

 View more versions of this product

Catalog No. AA13802A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-26-1
    Physical Form Crystals
    Molecular Formula H4AlNO8S2·12H2O
    Formula Weight 453.33
    MDL Number MFCD00051042
    Synonym Ammonium alum
    Chemical Name or Material Aluminum ammonium sulfate dodecahydrate
    Grade Reagent Grade

    Specifications

    Physical Form Crystals
    Formula Weight 453.33
    Quantity 2kg
    Color White
    EINECS Number 232-055-3
    CAS 7784-26-1
    Molecular Formula H4AlNO8S2·12H2O
    MDL Number MFCD00051042
    Synonym Ammonium alum
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Aluminum ammonium sulfate dodecahydrate
    Grade Reagent Grade

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit