Learn More
Aluminum ammonium sulfate dodecahydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper platingAluminum ammonium sulfate dodecahydrate acts as a crystalline double sulfate. It is used in water purification, in vegetable glues, in porcelain cements, in deodorant, in tanning and in fireproofing textiles. It serves as an ingredient in animal repellant sprays. Further, it is used in the preparation of Alum (Al(OH)3) gel. In addition, it is used in cosmetic formulation, fertilizers, food preservation and in the heating system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 93°C to 94°C |
Color | White |
Physical Form | Crystals |
Quantity | 500 g |
Solubility Information | Soluble in water and glycerol. Insoluble in alcohol. |
Formula Weight | 453.33 (237.14 Anhydrous) |
Grade | Reagent |
Density | 1.65 |
Chemical Name or Material | Aluminum ammonium sulfate dodecahydrate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 232-055-3 |
RTECSNumber | WS5640010 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.