Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol

Catalog No. p-7050363
Click to view available options
Quantity:
25 mL
100 mL

CAS: 23275-27-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23275-27-6
MDL Number MFCD00143586
Synonym acmc-20altq, calcium alumane pentakis propan-2-olate

Specifications

Boiling Point 230°C to 240°C (4.0 mmHg)
Odor Alcohol-like
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 566.3
Percent Purity 99.8%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Aluminum calcium isopropoxide
Hazard Category H225-H302-H314-H335-H336
Hazard Statement GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.