Learn More
Aluminum chloride, anhydrous, 99.985% (metals basis)
Description
Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 190° (sublimation) |
Color | Gray to Yellow |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.985% (Metal basis) |
Solubility Information | Soluble in water,hydrochloric acid and ethanol. Slightly soluble in benzene. |
Formula Weight | 133.34 |
Odor | Pungent |
Sensitivity | Moisture sensitive |
Appearance | Yellow to gray |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 231-208-1 |
RTECSNumber | BD0525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.